N-(4-Bromophenyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-[N-(4-Bromophenyl)anilino]naphthalene, N-(4-Bromophenyl)-N-phenylnaphthalen-1-amine

B4299-200MG B4299-1G
TCIAB4299-200MGEA 610 NOK
TCIAB4299-200MG TCIAB4299-1G
N-(4-Bromophenyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)
N-(4-Bromophenyl)-N-phenyl-1-naphthylamine
Formel: C₂₂H₁₆BrN
MW: 374,28 g/mol
MDL nummer: MFCD11046471
CAS-nummer: 138310-84-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR