Bis(2-ethylhexyl)isophthalate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Isophthalic Acid Bis(2-ethylhexyl) Ester
B4302-25G
B4302-500G
TCIAB4302-25GEA
220
NOK
TCIAB4302-25G
TCIAB4302-500G
Bis(2-ethylhexyl)isophthalate ≥98.0% (by GC)
Bis(2-ethylhexyl)isophthalate
Formel:
C₂₄H₃₈O₄ MW: 390,56 g/mol Kokepunkt: 211°C Flash punkt: 233°C |
MDL nummer:
MFCD00152284 CAS-nummer: 137-89-3 EINECS: 205-308-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...