4-tert-Butylphenylacetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(4-(tert-Butyl)phenyl)acetic acid

B4307-5G B4307-25G
TCIAB4307-5GEA 555 NOK
TCIAB4307-5G TCIAB4307-25G
4-tert-Butylphenylacetic acid ≥98.0% (by GC, titration analysis)
4-tert-Butylphenylacetic acid
Formel: C₁₂H₁₆O₂
MW: 192,26 g/mol
MDL nummer: MFCD00082593
CAS-nummer: 32857-63-9
EINECS: 251-264-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR