4-Bromo-3-fluorobenzaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
B4319-5G
B4319-25G
TCIAB4319-5GEA
635
NOK
TCIAB4319-5G
TCIAB4319-25G
4-Bromo-3-fluorobenzaldehyde ≥98.0% (by GC)
4-Bromo-3-fluorobenzaldehyde
Formel:
C₇H₄BrFO MW: 203,01 g/mol |
MDL nummer:
MFCD03095000 CAS-nummer: 133059-43-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...