Bis(3,5-dimethylphenyl)phosphine oxide ≥96.0% (by HPLC)

Leverandør: TCI
Warning

B4390-1G B4390-5G
TCIAB4390-1GEA 475 NOK
TCIAB4390-1G TCIAB4390-5G
Bis(3,5-dimethylphenyl)phosphine oxide ≥96.0% (by HPLC)
Bis(3,5-dimethylphenyl)phosphine oxide
Formel: C₁₆H₁₉OP
MDL nummer: MFCD08458884
CAS-nummer: 187344-92-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR