2-Bromo-9,9-dihexylfluorene ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Bromo-9,9-dihexyl-9H-fluorene

B4404-200MG B4404-1G
TCIAB4404-200MGEA 580 NOK
TCIAB4404-200MG TCIAB4404-1G
2-Bromo-9,9-dihexylfluorene ≥97.0% (by GC)
2-Bromo-9,9-dihexylfluorene
Formel: C₂₅H₃₃Br
MW: 413,44 g/mol
MDL nummer: MFCD14584649
CAS-nummer: 226070-05-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid to slightly cloudy liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR