2-Bromo-4-chloropyridine ≥98.0% (by GC)

Leverandør: TCI
Warning

B4407-1G B4407-5G
TCIAB4407-1GEA 350 NOK
TCIAB4407-1G TCIAB4407-5G
2-Bromo-4-chloropyridine ≥98.0% (by GC)
2-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
Kokepunkt: 85°C
MDL nummer: MFCD03840757
CAS-nummer: 22918-01-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 25.0 to 29.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR