4-Bromodibenzothiophene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4-Bromodibenzo[b,d]thiophene

B4449-1G B4449-5G
TCIAB4449-1GEA 340 NOK
TCIAB4449-1G TCIAB4449-5G
4-Bromodibenzothiophene ≥98.0% (by GC)
4-Bromodibenzothiophene
Formel: C₁₂H₇BrS
MW: 263,15 g/mol
MDL nummer: MFCD02683746
CAS-nummer: 97511-05-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR