5-Bromo-2-methox-6-methylypyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine
B4460-1G
B4460-5G
TCIAB4460-1GEA
220
NOK
TCIAB4460-1G
TCIAB4460-5G
5-Bromo-2-methox-6-methylypyridine ≥98.0% (by GC)
5-Bromo-2-methox-6-methylypyridine
Formel:
C₇H₈BrNO MW: 202,05 g/mol Kokepunkt: 86°C Flash punkt: 106°C |
MDL nummer:
MFCD03094943 CAS-nummer: 126717-59-7 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...