N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
B4478-1G
B4478-5G
TCIAB4478-1GEA
700
NOK
TCIAB4478-1G
TCIAB4478-5G
N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate
Formel:
C₁₄H₁₈N₄·₄HCl·2H₂O MW: 388,16 g/mol |
CAS-nummer:
1310544-60-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 219.0 to 223.0 deg-C |
Water | 7.0 to 10.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...