Bis(pentafluorophenyl)phenylphosphine ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Bis(perfluorophenyl)(phenyl)phosphine, SpectraSynQ 443

B4560-200MG B4560-1G
TCIAB4560-200MGEA 500 NOK
TCIAB4560-200MG TCIAB4560-1G
Bis(pentafluorophenyl)phenylphosphine ≥98.0% (by GC)
Bis(pentafluorophenyl)phenylphosphine
Formel: C₁₈H₅F₁₀P
MW: 442,2 g/mol
MDL nummer: MFCD00000291
CAS-nummer: 5074-71-5
EINECS: 225-780-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR