1-Bromo-2-ethynylbenzene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2'-Bromophenylacetylene, 2-Bromophenylacetylene

B4608-1G B4608-5G
TCIAB4608-1GEA 1375 NOK
TCIAB4608-1G TCIAB4608-5G
1-Bromo-2-ethynylbenzene ≥98.0% (by GC)
1-Bromo-2-ethynylbenzene
Formel: C₈H₅Br
MW: 181,03 g/mol
Kokepunkt: 94°C
Flash punkt: 63°C
MDL nummer: MFCD01863358
CAS-nummer: 766-46-1

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR