2-(tert-Butyldimethylsilyl)thiazole ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
B4654-1G
B4654-5G
TCIAB4654-1GEA
475
NOK
TCIAB4654-1G
TCIAB4654-5G
2-(tert-Butyldimethylsilyl)thiazole ≥97.0% (by GC)
2-(tert-Butyldimethylsilyl)thiazole
Formel:
C₉H₁₇NSSi |
MDL nummer:
MFCD10699169 CAS-nummer: 137382-38-8 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...