9,9-Bis(3,4-dihydroxyphenyl)fluorene ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
4,4'-(9H-Fluorene-9,9-diyl)bis(benzene-1,2-diol)
B4665-1G
B4665-5G
TCIAB4665-1GEA
1025
NOK
TCIAB4665-1G
TCIAB4665-5G
9,9-Bis(3,4-dihydroxyphenyl)fluorene ≥98.0% (by GC)
9,9-Bis(3,4-dihydroxyphenyl)fluorene
Formel:
C₂₅H₁₈O₄ MW: 382,42 g/mol |
CAS-nummer:
351521-78-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...