9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)
Leverandør: TCI
Synonymer:
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene Diglycidyl ether
B4666-1G
B4666-5G
TCIAB4666-1GEA
1425
NOK
TCIAB4666-1G
TCIAB4666-5G
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene
Formel:
C₃₃H₃₀O₄ MW: 490,60 g/mol |
MDL nummer:
MFCD22494984 CAS-nummer: 114205-89-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...