4-(6-Amino-3-pyridyl)-1-boc-piperazine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester

B4669-200MG B4669-1G
TCIAB4669-200MGEA 580 NOK
TCIAB4669-200MG TCIAB4669-1G
4-(6-Amino-3-pyridyl)-1-boc-piperazine ≥98.0% (by HPLC, titration analysis)
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Formel: C₁₄H₂₂N₂O₄
MW: 278,35 g/mol
MDL nummer: MFCD11594962
CAS-nummer: 571188-59-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR