4-Bromo-2-(1H-pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
B4743-1G
B4743-5G
TCIAB4743-1GEA
270
NOK
TCIAB4743-1G
TCIAB4743-5G
4-Bromo-2-(1H-pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
4-Bromo-2-(1H-pyrazol-3-yl)phenol
Formel:
C₉H₇BrN₂O MW: 239,08 g/mol |
MDL nummer:
MFCD03094026 CAS-nummer: 99067-15-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Purple powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...