1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, tert-Butyl 3-Fluoro-4-oxo-1-piperidinecarboxylate, tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate, 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, 3-Fluoro-4-oxo-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-fluoro-4-piperidone

B4858-200MG B4858-1G
TCIAB4858-200MGEA 1025 NOK
TCIAB4858-200MG TCIAB4858-1G
1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)
1-Boc-3-fluoro-4-piperidone
Formel: C₁₀H₁₆FNO₃
MW: 217,24 g/mol
MDL nummer: MFCD10566554
CAS-nummer: 211108-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8de80e375979a577d85884a1b61b890c4154f2a0&vid=8de80e375979a577d85884a1b61b890c4154f2a0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR