5-Bromo-2-chloronicotinonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 5-Bromo-2-chloro-3-cyanopyridine

B4870-1G B4870-5G
TCIAB4870-1GEA 620 NOK
TCIAB4870-1G TCIAB4870-5G
5-Bromo-2-chloronicotinonitrile ≥98.0% (by GC)
5-Bromo-2-chloronicotinonitrile
Formel: C₆H₂BrClN₂
MDL nummer: MFCD09801046
CAS-nummer: 405224-23-9
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR