4-(4-Bromophenyl)dibenzofuran ≥98.0% (by GC)

Leverandør: TCI

B4884-1G B4884-5G
TCIAB4884-1GEA 1325 NOK
TCIAB4884-1G TCIAB4884-5G
4-(4-Bromophenyl)dibenzofuran ≥98.0% (by GC)
4-(4-Bromophenyl)dibenzofuran
Formel: C₁₈H₁₁BrO
MW: 323,19 g/mol
MDL nummer: MFCD19441306
CAS-nummer: 955959-84-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR