3-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
3-Bromo-5'-phenyl-m-terphenyl, 5'-(3-Bromophenyl)-1,1':3',1''-terphenyl, 5'-(3-Bromophenyl)-m-terphenyl
B4888-1G
B4888-5G
TCIAB4888-1GEA
985
NOK
TCIAB4888-1G
TCIAB4888-5G
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formel:
C₂₄H₁₇Br MW: 385,30 g/mol |
MDL nummer:
MFCD28134530 CAS-nummer: 1233200-57-1 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...