2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

B4894-1G B4894-5G
TCIAB4894-1GEA 915 NOK
TCIAB4894-1G TCIAB4894-5G
2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)
2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Formel: C₂₁H₁₄BrN₃
MW: 388,27 g/mol
MDL nummer: MFCD22200060
CAS-nummer: 864377-31-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR