6-Bromoquinolin-4-ol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Bromo-4-quinolinol, 6-Bromo-4-hydroxyquinoline
B4899-1G
B4899-5G
TCIAB4899-1GEA
945
NOK
TCIAB4899-1G
TCIAB4899-5G
6-Bromoquinolin-4-ol ≥98.0% (by GC, titration analysis)
6-Bromoquinolin-4-ol
Formel:
C₉H₆BrNO MW: 224,0540 g/mol Smeltepunkt: 283°C |
MDL nummer:
MFCD00272435 CAS-nummer: 145369-94-4 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...