2-Bromo-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
B4939-1G
B4939-5G
TCIAB4939-1GEA
960
NOK
TCIAB4939-1G
TCIAB4939-5G
2-Bromo-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
2-Bromo-4,6-diphenyl-1,3,5-triazine
Formel:
C₁₅H₁₀BrN₃ MW: 312,17 g/mol |
MDL nummer:
MFCD10687147 CAS-nummer: 80984-79-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...