4-Bromobutyl acetate ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
Acetic acid 4-bromobutyl ester
B4958-5G
B4958-25G
TCIAB4958-5GEA
310
NOK
TCIAB4958-5G
TCIAB4958-25G
4-Bromobutyl acetate ≥97.0% (by GC)
4-Bromobutyl acetate
Formel:
C₆H₁₁BrO₂ MW: 195,06 g/mol Kokepunkt: 211°C |
MDL nummer:
MFCD00000263 CAS-nummer: 4753-59-7 EINECS: 225-277-7 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...