6-Bromo-3-cyanochromone ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile, 6-Bromochromone-3-carbonitrile, 6-Bromo-4-oxo-4H-chromene-3-carbonitrile

B4968-1G B4968-5G
TCIAB4968-1GEA 700 NOK
TCIAB4968-1G TCIAB4968-5G
6-Bromo-3-cyanochromone ≥98.0% (by GC)
6-Bromo-3-cyanochromone
Formel: C₁₀H₄BrNO₂
MW: 250,483 g/mol
MDL nummer: MFCD00191842
CAS-nummer: 52817-13-7
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c7878d9b3f6c9c35938d4dcf906b7eb7d743c7d&vid=5c7878d9b3f6c9c35938d4dcf906b7eb7d743c7d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR