2-(4-Bromophenyl)benzoxazole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-(4-Bromophenyl)benzo[d]oxazole

B5017-1G B5017-5G
TCIAB5017-1GEA 785 NOK
TCIAB5017-1G TCIAB5017-5G
2-(4-Bromophenyl)benzoxazole ≥98.0% (by GC)
2-(4-Bromophenyl)benzoxazole
Formel: C₁₃H₈BrNO
MW: 274,12 g/mol
MDL nummer: MFCD00168898
CAS-nummer: 3164-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3e911fc78490ee6a6c7e8b475ef025fedfa5d8a&vid=d3e911fc78490ee6a6c7e8b475ef025fedfa5d8a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR