9,10-Bis(4-methoxyphenyl)anthracene ≥98.0% (by GC)
Leverandør: TCI
B5022-200MG
B5022-1G
TCIAB5022-200MGEA
745
NOK
TCIAB5022-200MG
TCIAB5022-1G
9,10-Bis(4-methoxyphenyl)anthracene ≥98.0% (by GC)
9,10-Bis(4-methoxyphenyl)anthracene
Formel:
C₂₈H₂₂O₂ MW: 390.48 g/mol |
MDL nummer:
MFCD00009936 CAS-nummer: 24672-76-2 |
|
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...