3-Bromo-9-(p-tolyl)-9H-carbazole ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
B5052-1G
B5052-5G
TCIAB5052-1GEA
755
NOK
TCIAB5052-1G
TCIAB5052-5G
3-Bromo-9-(p-tolyl)-9H-carbazole ≥97.0% (by GC)
3-Bromo-9-(p-tolyl)-9H-carbazole
Formel:
C₁₉H₁₄BrN MW: 336,23 g/mol |
MDL nummer:
MFCD16658912 CAS-nummer: 731016-44-7 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...