2-(4-Bromophenyl)thiophene ≥97.0% (by GC)
Leverandør: TCI
B5106-1G
B5106-5G
TCIAB5106-1GEA
1775
NOK
TCIAB5106-1G
TCIAB5106-5G
2-(4-Bromophenyl)thiophene ≥97.0% (by GC)
2-(4-Bromophenyl)thiophene
Formel:
C₁₀H₇BrS MW: 239,13 g/mol |
MDL nummer:
MFCD05664340 CAS-nummer: 40133-22-0 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...