2-Bromo-9,9-diethylfluorene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Bromo-9,9-diethyl-9H-fluorene
B5126-1G
B5126-5G
TCIAB5126-1GEA
890
NOK
TCIAB5126-1G
TCIAB5126-5G
2-Bromo-9,9-diethylfluorene ≥98.0% (by GC)
2-Bromo-9,9-diethylfluorene
Formel:
C₁₇H₁₇Br MW: 301,23 g/mol Kokepunkt: 163°C |
MDL nummer:
MFCD09909863 CAS-nummer: 287493-15-6 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...