3-Bromo-2-fluorobenzylalkohol ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(3-Bromo-2-fluorophenyl)methanol, 3-Bromo-2-fluorobenzyl alcohol, 1-Bromo-2-fluoro-3-(hydroxymethyl)benzene
B5143-1G
B5143-5G
TCIAB5143-1GEA
365
NOK
TCIAB5143-1G
TCIAB5143-5G
3-Bromo-2-fluorobenzylalkohol ≥98.0% (by GC)
3-Bromo-2-fluorobenzylalkohol
Formel:
C₇H₆BrFO MW: 205,0200 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD09842441 CAS-nummer: 261723-32-4 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...