Barium bis(trifluoromethylsulphonyl)imide ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Barium triflimide, Bis(trifluoromethanesulphonyl)imide barium(II) salt, Barium bis(trifluoromethanesulphonimide), Barium(II) triflimide, Barium(II) Bis(trifluoromethanesulphonyl)imide

B5171-1G B5171-5G
TCIAB5171-1GEA 1050 NOK
TCIAB5171-1G TCIAB5171-5G
Barium bis(trifluoromethylsulphonyl)imide ≥99.0% (by titrimetric analysis)
Barium bis(trifluoromethylsulphonyl)imide
Formel: C₄BaF₁₂N₂O₈S₄
MW: 697,60 g/mol
MDL nummer: MFCD22200529
CAS-nummer: 168106-22-7
FN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9809ef518a4cbb1f2d01b432b69c720405ab01ea&vid=9809ef518a4cbb1f2d01b432b69c720405ab01ea&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR