5-Bromo-2-methoxyphenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 5-Bromo-2-methoxybenzeneboronic acid

B5173-1G B5173-5G
TCIAB5173-1GEA 325 NOK
TCIAB5173-1G TCIAB5173-5G
5-Bromo-2-methoxyphenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
5-Bromo-2-methoxyphenylboronic acid
Formel: C₇H₈BBrO₃
MW: 230,85 g/mol
Smeltepunkt: 131°C
MDL nummer: MFCD00093409
CAS-nummer: 89694-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaf59f4f0bb82c7310e10014e2486e95713dcd04&vid=eaf59f4f0bb82c7310e10014e2486e95713dcd04&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 95.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR