4-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
(4-Bromo-2-methylphenyl)methanol
B5231-1G
B5231-5G
TCIAB5231-1GEA
595
NOK
TCIAB5231-1G
TCIAB5231-5G
4-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)
4-Bromo-2-methylbenzyl alcohol
Formel:
C₈H₉BrO MW: 201,06 g/mol |
MDL nummer:
MFCD11847398 CAS-nummer: 17100-58-2 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...