2-Bromo-4,6-dimethylpyridine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
B5254-1G
B5254-5G
TCIAB5254-1GEA
1525
NOK
TCIAB5254-1G
TCIAB5254-5G
2-Bromo-4,6-dimethylpyridine ≥98.0% (by titrimetric analysis)
2-Bromo-4,6-dimethylpyridine
Formel:
C₇H₈BrN MW: 186,05 g/mol Kokepunkt: 68°C |
MDL nummer:
MFCD00082591 CAS-nummer: 4926-26-5 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...