Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde ≥95.0% (by GC)

Leverandør: TCI

B5279-200MG B5279-1G
TCIAB5279-200MGEA 1000 NOK
TCIAB5279-200MG TCIAB5279-1G
Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde ≥95.0% (by GC)
Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde
Formel: C₉H₁₂O
MW: 136,19 g/mol
Kokepunkt: 73°C
Smeltepunkt: 13°C
Tetthet: 1,04
Lagringstemperatur: Kjøleskap
CAS-nummer: 38259-00-6

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR