3-Bromo-5-nitropyridin-4-ol ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Bromo-4-hydroxy-5-nitropyridine, 3-Bromo-5-nitro-4-pyridinol
B5287-1G
B5287-5G
TCIAB5287-1GEA
960
NOK
TCIAB5287-1G
TCIAB5287-5G
3-Bromo-5-nitropyridin-4-ol ≥98.0% (by titrimetric analysis)
3-Bromo-5-nitropyridin-4-ol
Formel:
C₅H₃BrN₂O₃ MW: 219 g/mol |
MDL nummer:
MFCD08692339 CAS-nummer: 31872-65-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...