α,3-Dibromo-5-fluorotoluene ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 3-Bromo-5-fluorobenzyl bromide, 1-Bromo-3-(bromomethyl)-5-fluorobenzene

B5338-5G B5338-25G
TCIAB5338-5GEA 1100 NOK
TCIAB5338-5G TCIAB5338-25G
α,3-Dibromo-5-fluorotoluene ≥98.0% (by GC)
α,3-Dibromo-5-fluorotoluene
Formel: C₇H₅Br₂F
MW: 267,92 g/mol
MDL nummer: MFCD09258959
CAS-nummer: 216755-57-6
FN: 3261
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR