3-[(tert-Butoxycarbonyl)amino]thiophene-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-(Boc-amino)thiophene-2-carboxylic acid, 3-(tert-Butoxycarbonylamino)thiophene-2-carboxylic acid, 3-(N-Boc-amino)thiophene-2-carboxylic acid

B5341-1G B5341-5G
TCIAB5341-1GEA 915 NOK
TCIAB5341-1G TCIAB5341-5G
3-[(tert-Butoxycarbonyl)amino]thiophene-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3-[(tert-Butoxycarbonyl)amino]thiophene-2-carboxylic acid
Formel: C₁₀H₁₃NO₄S
MW: 243,28 g/mol
MDL nummer: MFCD02682396
CAS-nummer: 101537-64-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR