(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate HCl, Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate hydrochloride, (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic acid Benzyl ester hydrochloride
B5344-1G
B5344-5G
TCIAB5344-1GEA
635
NOK
TCIAB5344-1G
TCIAB5344-5G
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥98.0% (by HPLC, titration analysis)
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Formel:
C₁₅H₂₀ClNO₂ MW: 281,7800 g/mol |
MDL nummer:
MFCD04974076 CAS-nummer: 87269-87-2 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | -28.0 to -32.0 deg (C=1, DMSO) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...