2-Bromoallyl alcohol ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Bromo-2-propen-1-ol, 2-Bromoprop-2-en-1-ol

B5347-1G B5347-5G
TCIAB5347-1GEA 1300 NOK
TCIAB5347-1G TCIAB5347-5G
2-Bromoallyl alcohol ≥97.0% (by GC)
2-Bromoallyl alcohol
Formel: C₃H₅BrO
MW: 136,98 g/mol
Kokepunkt: 150°C
Flash punkt: 76°C
MDL nummer: MFCD09265077
CAS-nummer: 598-19-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc422420fc1f7fcd58f4803750f812b7663f618&vid=ffc422420fc1f7fcd58f4803750f812b7663f618&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR