3-Bromo-2-fluorobenzaldehyde ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Bromo-2-fluorobenzenecarbaldehyde

B5363-5G B5363-25G
TCIAB5363-5GEA 810 NOK
TCIAB5363-5G TCIAB5363-25G
3-Bromo-2-fluorobenzaldehyde ≥98.0% (by GC)
3-Bromo-2-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL nummer: MFCD09027089
CAS-nummer: 149947-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc4b391bfb24c8a27c86425541b69c76f921206a&vid=cc4b391bfb24c8a27c86425541b69c76f921206a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR