4-(4-Bromophenyl)dibenzothiophene ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
4-(4-Bromophenyl)dibenzo[b,d]thiophene
B5384-200MG
B5384-1G
TCIAB5384-200MGEA
500
NOK
TCIAB5384-200MG
TCIAB5384-1G
4-(4-Bromophenyl)dibenzothiophene ≥98.0% (by GC)
4-(4-Bromophenyl)dibenzothiophene
Formel:
C₁₈H₁₁BrS MW: 339,25 g/mol |
MDL nummer:
MFCD19441307 CAS-nummer: 530402-77-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...