1-Benzyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
1-Benzyl-1H-pyrazole-4-carboxaldehyde
B5404-1G
B5404-200MG
TCIAB5404-1GEA
1625
NOK
TCIAB5404-1G
TCIAB5404-200MG
1-Benzyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
1-Benzyl-1H-pyrazole-4-carbaldehyde
Formel:
C₁₁H₁₀N₂O MW: 186,2099 g/mol Kokepunkt: 144°C |
MDL nummer:
MFCD02179567 CAS-nummer: 63874-95-3 |
Spesifikasjon Testresultater
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...