BMeS-p-A ≥95.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 1,4-Diamino-2,5-bis(methylsulphonyl)benzene, 2,5-Bis(methylsulphonyl)-1,4-phenylenediamine

B5419-200MG B5419-1G
TCIAB5419-200MGEA 6625 NOK
TCIAB5419-200MG TCIAB5419-1G
BMeS-p-A ≥95.0% (by HPLC)
BMeS-p-A
Formel: C₈H₁₂N₂O₄S₂
MW: 264,31 g/mol
CAS-nummer: 1678513-97-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0e33d4184ac7acc8b87b94f3074c65e9b4dba4f&vid=c0e33d4184ac7acc8b87b94f3074c65e9b4dba4f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 387.0 to 392.0 nm (DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR