3-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
B5436-200MG
B5436-1G
TCIAB5436-200MGEA
715
NOK
TCIAB5436-200MG
TCIAB5436-1G
3-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
3-Bromo-9,9-dimethyl-9H-fluorene
Formel:
C₁₅H₁₃Br MW: 273,17 g/mol |
MDL nummer:
MFCD28955974 CAS-nummer: 1190360-23-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...