Bis(tetrabutylammonium) dihydrogen Pyrophosphate ≥96.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
B5440-200MG
B5440-1G
TCIAB5440-200MGEA
2250
NOK
TCIAB5440-200MG
TCIAB5440-1G
Bis(tetrabutylammonium) dihydrogen Pyrophosphate ≥96.0% (by titrimetric analysis)
Bis(tetrabutylammonium) dihydrogen Pyrophosphate
Formel:
C₃₂H₇₄N₂O₇P₂ MW: 660,90 g/mol Lagringstemperatur: Fryseskap |
CAS-nummer:
857447-79-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...