1-Butyl-1-methylpyrrolidiniumdicyanamide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
B5453-5G
B5453-25G
TCIAB5453-5GEA
770
NOK
TCIAB5453-5G
TCIAB5453-25G
1-Butyl-1-methylpyrrolidiniumdicyanamide ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidiniumdicyanamide
Formel:
C₁₁H₂₀N₄ MW: 208,31 g/mol |
MDL nummer:
MFCD08458923 CAS-nummer: 370865-80-8 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...