4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 4,4'-Bis(trifluoromethyl)-2,2'-bipyridine

B5463-200MG B5463-1G
TCIAB5463-200MGEA 960 NOK
TCIAB5463-200MG TCIAB5463-1G
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl
Formel: C₁₂H₆F₆N₂
MW: 292,18 g/mol
MDL nummer: MFCD00233878
CAS-nummer: 142946-79-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR